CaptureSelect™ FcXP Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ FcXP Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8103712011 Assay x 96-well Plate
81037121010 Assay x 96-well Plate
Catalog number 810371201
Price (USD)
1,250.00
Each
Quantity:
1 Assay x 96-well Plate
Price (USD)
1,250.00
Each

CaptureSelect FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix. The FcXP affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FcXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Limit1 ng/mL
FormatIndividual components
Ligand TypeCaptureSelect FcXP Affinity Ligand
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay x 96-well Plate
Shipping ConditionDry Ice
Sufficient For1 x 96-well plate
TargetCaptureSelect FcXP Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Unit SizeEach
Contents & Storage
Store at -20°C