CaptureSelect™ AAT-XL Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ AAT-XL Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ AAT-XL Ligand Leakage ELISA is designed to detect possible leached ligand of alpha-1 anti-trypsin (AAT) purified using CaptureSelect AAT-XL Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8103832011 Assay x 96-well plate
81038321010 Assay x 96-well Plate
Catalog number 810383201
Price (USD)
1,250.00
Each
Quantity:
1 Assay x 96-well plate
Price (USD)
1,250.00
Each

CaptureSelect AAT-XL Ligand Leakage ELISA is designed to detect possible leached ligand of alpha-1 anti-trypsin (AAT) purified using CaptureSelect AAT-XL Affinity Matrix. The AAT-XL affinity ligand is a 13-kD recombinant single-domain antibody fragment (VHH affinity ligand) expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of ligand can leach from the chromatography support and co-elute with the target protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand concentrations greater than or equal to 1 ng/mL. The CaptureSelect AAT-XL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications
Detection Limit1 ng/mL
For Use With (Application)Ligand Leakage ELISA assay
FormatIndividual components
Ligand TypeCaptureSelect Human Alpha-1 Antitrypsin (AAT) Affinity Ligand
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay x 96-well plate
Shipping ConditionDry Ice
TargetTarget-specific ELISA reagents
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Unit SizeEach
Contents & Storage
Store at -20°C