Which syringe(s) do you recommend using with your mass spectrometry calibration solutions?
We recommend using a 500 µl Hamilton Syringe (Cat. No. 81265) for direct infusion of calibration solutions. The syringe and tubing can be cleaned with LC-MS grade 50% MeOH/50% water between uses and should be stored dry. Avoid storing the syringe or tubing with the calibration solution.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.