Orion™ ROSS Ultra™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Thermo Scientific™

Orion™ ROSS Ultra™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode

Measure flat, soft, moist, semi-solid or small volume samples with epoxy body flat surface pH electrodes.
Have Questions?
Catalog NumberConnector Types
8135BNUWPBNC (Waterproof)
Catalog number 8135BNUWP
Price (USD)
762.30
Online exclusive
847.00
Save 84.70
Each
Connector Types:
BNC (Waterproof)
Price (USD)
762.30
Online exclusive
847.00
Save 84.70
Each
Thermo Scientific™ Orion™ ROSS Ultra Flat Surface Electrodes have a uniquely designed, flat sensing bulb that allow for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates.
 

Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonomous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 24 months from date of purchase
Specifications
DescriptionUltra pH Electrode
ElectrodeFlat surface tip epoxy body combination electrode; 20 x 12mm
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter
Internal ReferenceROSS
Length (English) Cable39 in.
Length (Metric)1 m
Precision0.01
Temperature (English) Operating32°F to 212°F
Warranty2 Years
Connector TypesBNC (Waterproof)
JunctionGlass Fiber
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 100°C
TypepH Electrode
pH Range0 to 14
Unit SizeEach