Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezer, 12.7 cu. ft. (359.6L) capacity, 230V/50Hz, 12A - FAQs
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezer, 12.7 cu. ft. (359.6L) capacity, 230V/50Hz, 12A - FAQs
View additional product information for Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (813CV)