Orion™ ROSS Ultra™ Refillable Epoxy Bodied Combination pH Electrode
Orion™ ROSS Ultra™ Refillable Epoxy Bodied Combination pH Electrode
Orion™ ROSS Ultra™ Refillable Epoxy Bodied Combination pH Electrode
Thermo Scientific™

Orion™ ROSS Ultra™ Refillable Epoxy Bodied Combination pH Electrode

Confidently maneuver and measure in the field or on the plant floor with Orion refillable epoxy body electrodes.
Have Questions?
Catalog NumberConnector Types
8156BNUWPBNC (Waterproof)
Catalog number 8156BNUWP
Price (USD)
623.70
Online exclusive
693.00
Save 69.30
Each
Connector Types:
BNC (Waterproof)
Price (USD)
623.70
Online exclusive
693.00
Save 69.30
Each
Thermo Scientific™ Orion™ ROSS Refillable Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. They are available in epoxy bodies for ruggedness and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Orion ROSS Ultra Refillable electrodes come with a two-year warranty.

Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonomous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required

  • Electrode fill solution
  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
  • electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
  • storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

24 months from date of purchase

Specifications
DescriptionGeneral purpose epoxy body combination electrode; 120 x 12mm
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter, BNC connector
Internal ReferenceROSS
Length (English) Cable39 in.
Length (Metric) Cable1 m
Precision0.01
Temperature (English) Operating32°F to 212°F
Warranty2 Years
Connector TypesBNC (Waterproof)
JunctionGlass Fiber
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 100°C
TypepH Electrode
pH Range0 to 14
Unit SizeEach