Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode
Thermo Scientific™

Orion™ ROSS™ Sure-Flow™ Epoxy Bodied Combination pH Electrode

Attain stable, drift-free measurements of challenging samples using a sure-flow electrode with its unique free-flowing liquid-to-liquid junction.
Have Questions?
Catalog NumberConnector Types
8165BNWPBNC (Waterproof)
Catalog number 8165BNWP
Price (USD)
654.30
Online exclusive
727.00
Save 72.70
Each
Connector Types:
BNC (Waterproof)
Price (USD)
654.30
Online exclusive
727.00
Save 72.70
Each
Thermo Scientific™ Orion™ Sure-Flow™ electrodes are built with a unique free-flowing liquid-to-liquid junction that simplifies measurements of dirty or viscous samples by helping to prevent clogging, making them easy to clean. They are available in epoxy bodies for ruggedness and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
 

Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionWith waterproof connector
Diameter (Metric) Cap16 mm
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter, BNC connector
IncludesDetachable bulb guard, 2 x 60mL bottles of fill solution
Internal ReferenceROSS
Length (English) Cable39 in.
Length (Metric) Cable1 m
Precision0.01
Temperature (English) Operating32°F to 212°F
Connector TypesBNC (Waterproof)
JunctionSure-Flow
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 100°C
TypeCombination pH Electrode, epoxy body, BNC waterproof connector
pH Range0 to 14
Unit SizeEach