Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode

Measure small volumes down to 15 μL in 384 well plates with micro pH electrodes.
Have Questions?
Catalog NumberConnector Types
8220BNWPBNC (Waterproof)
Catalog number 8220BNWP
Price (USD)
772.20
Online exclusive
858.00
Save 85.80
Each
Connector Types:
BNC (Waterproof)
Price (USD)
772.20
Online exclusive
858.00
Save 85.80
Each
Thermo Scientific™ Orion™ ROSS PerpHecT™ Micro Combination Electrodes are best suited for use in biological samples, where they are much more versatile than silver chloride electrodes. When used in conjunction with our Orion LogR technology, the glass bulb of the electrode will measure both pH and temperature at the same location, which allows for extremely accurate measurement in small volumes.
 

Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required

  • Electrode fill solution
  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
  • electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
  • storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionCombination pH Micro Electrode
Diameter (Metric) Cap3 mm
DriftLess than 0.002 pH units/day
ElectrodeGlass double junction; Refillable
For Use With (Equipment)Standard pH Meter
Length (Metric)155 mm
Length (Metric) Cable1 m
Min. Immersion4.5mm
Storage RequirementsTo ensure a quick response and free-flowing liquid junction the sensing element and reference junction must not be allowed to dry out. Short-term Storage (up to one week): Soak electrode in ROSS pH Electrode Storage Solution, Orion 810001 or use pH Electrode Storage Solution, Orion 910001. To minimize the chance of breakage the 8220BNWP should be attached to an electrode stand and suspended in a beaker that contains pH electrode storage solution. The electrode should not touch the sides or bottom of the beaker. Long-term Storage (more than one week): The reference chamber should be filled and the filling hole securely covered. Cover the sensing element and reference Junction with the protective cap/sleeve containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Connector TypesBNC (Waterproof)
JunctionGlass Double
MaterialGlass
Temperature (Metric) Operating0°C to 100°C
TypeCombination pH Micro Electrode
pH Range0 to 14
Unit SizeEach

Frequently asked questions (FAQs)

What is the warranty period for the Orion ROSS PerpHecT Micro Glass Bodied Combination pH Electrode?

The warranty period for the Orion ROSS PerpHecT Micro Glass Bodied Combination pH Electrode is 12 months from date of purchase.