Industrial Coatings FT-IR Spectral Library
Thermo Scientific™

Industrial Coatings FT-IR Spectral Library

Thermo Scientific™ Industrial Coatings FT-IR Spectral Library is a valuable resource for commercial and industrial laboratories.
Have Questions?
Catalog NumberType
834-010600Industrial Coatings FT-IR Spectral Library
Catalog number 834-010600
Price (MXN)
-
Type:
Industrial Coatings FT-IR Spectral Library

Thermo Scientific™ Industrial Coatings FT-IR Spectral Library is a valuable resource for commercial and industrial laboratories.

This library includes a wide-range of spectra of monomers, polymers, plasticizers, extenders, solvents, pigments and other additives.
  • Contains 1,961 spectra listed by their trade names
  • Data format: High Resolution
Specifications
For Use With (Application)FT-IR
TypeIndustrial Coatings FT-IR Spectral Library
Unit SizeEach