GENESYS Smart QC software
GENESYS Smart QC software
GENESYS Smart QC software
GENESYS Smart QC software
GENESYS Smart QC software
GENESYS Smart QC software
GENESYS Smart QC software
GENESYS Smart QC software
Thermo Scientific™
GENESYS Smart QC software
PC-controlled-method development with local control method deployment for UV-Vis analysis. Ideal for quality control settings, especially in ingredient management.
Have Questions?
Catalog NumberType
837-004500GENESYS Smart QC software
Catalog number 837-004500
Price (CLP)
-
Type:
GENESYS Smart QC software
Thermo Scientific™ GENESYS™ Smart QC Software can make your UV-Vis analysis smoother and easier. GENESYS Smart QC software provides a simple way to pass judgement on a sample being tested by UV-Vis spectroscopy, delivering a straightforward functionality not readily available from other manufacturers. With GENESYS Smart QC software, UV-Vis analysis methods can be customized to deliver simple pass/fail assessments based on your specific guidelines. This limits the risk of misinterpreting data and speeds the time to action by streamlining decision-making points for users at all levels . The size, simplicity, and cost-effective operation of a UV-Vis spectrophotometer and the range of tests available make it an ideal instrument for industrial QA/QC labs, especially those performing food and beverage analysis.
The GENESYS Smart QC software package takes a twofold approach: It utilizes PC-controlled method development, followed by locally controlled method deployment on the desired analytical instrument.
  • An analyst can use a personal computer (PC) to custom design a set of equations and designate how the data obtained on the local spectrophotometer should be interpreted.
  • Local control is intended to be as simple as possible, employing a straightforward, simple interface to limit the amount spectral interpretation required of the user.
The customized method can be transferred from a research director’s PC to a GENESYS UV-Vis Spectrophotometer in the lab, which allows the local operations to be performed by lab technicians regardless of familiarity with UV-Vis analytical methods. A typical result of the pre-programmed method could be as basic as a PASS/FAIL readout.
  • Method development is ideal for setting quality control parameters, especially for ingredient control
  • GENESYS Smart QC software can be used anywhere a simple instrument read-out with limited interpretation is required
Specifications
DescriptionGENESYS Smart QC software
TypeGENESYS Smart QC software
Unit SizeEach