Search
Search

| Número do catálogo | For Use With (Equipment) | Type |
|---|---|---|
| 840-307400 | GENESYS 150, GENESYS 180, Evolution One Plus, and Evolution Pro spectrophtometers | Tip for Dip Probe, 10 mm |
| 840-305000 | GENESYS 150, GENESYS 180, Evolution One Plus, and Evolution Pro spectrophtometers | Fiber Optic Dip Probe, 10 mm |
| 840-305700 | GENESYS 150/180 Spectrophotometer | Fiber Optic Dip Probe System |
| 840-305800 | GENESYS 150/180 Spectrophotometer | Fiber Optic Coupler |