NanoDrop™ Eight Tube Rack
NanoDrop™ Eight Tube Rack
NanoDrop™ Eight Tube Rack
NanoDrop™ Eight Tube Rack
The Thermo Scientific™ NanoDrop™ Eight Tube Rack allows you to place samples in tubes on the instrument LED board to help you keep track of samples measured.
Have Questions?
Catalog NumberFor Use With (Equipment)
840-343700NanoDrop Eight Spectrophotometers
Catalog number 840-343700
Price (BRL)
454,78
Each
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Price (BRL)
454,78
Each
The Thermo Scientific™ NanoDrop™ Eight Tube Rack allows you to place samples in tubes on the instrument LED board to help you keep track of samples measured.
This single tube rack allows users to keep track of sample measurements for samples stored in 0.5 mL and 1.5 mL-sized tubes.
Specifications
DescriptionNanoDrop Eight Tube Rack
For Use With (Equipment)NanoDrop Eight Spectrophotometers
Unit SizeEach
Frequently asked questions (FAQs)
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.