96 Well UV Transparent Plate, Pack of 10
Thermo Scientific™
96 Well UV Transparent Plate, Pack of 10
Perform DNA and protein quantitation in 260nm and 280nm range with Thermo Scientific™ Nunc™ 96-Well UV Microplates.
Have Questions?
Catalog NumberQuantity
8404Case of 40
Catalog number 8404
Price (USD)
810.00
Case of 40
Quantity:
Case of 40
Price (USD)
810.00
Case of 40
Perform DNA and protein quantitation in 260nm and 280nm range with Thermo Scientific™ Nunc™ 96-Well UV Microplates. The 96-Well format allows for a higher throughput of sample screening compared to cuvettes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionNunc 96 Well UV transparent plate packaged 10/40
LidWithout Lid
Packaging10/Pack, 40/Case
Product TypeMicroplate
QuantityCase of 40
Array8 x 12
ColorClear
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentUV
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 40