Sample Introduction Spares Kit for iCAP™ 7000 Series ICP-OES Systems
Thermo Scientific™
Sample Introduction Spares Kit for iCAP™ 7000 Series ICP-OES Systems
Simplify the ordering process to save time with the Thermo Scientific™ Sample Introduction Spares Kit for iCAP™ 7000 Series ICP-OES systems.
Have Questions?
Catalog NumberType
842312051751Sample Introduction Spares Kit
Catalog number 842312051751
Price (USD)
718.00
Each
Type:
Sample Introduction Spares Kit
Price (USD)
718.00
Each
Find the spares and consumables you need for sample introduction into Thermo Scientific™ iCAP™ 7000 Series ICP-OES systems. The Thermo Scientific™ Sample Introduction Spares Kit for iCAP 7000 Series ICP-OES systems simplifies the ordering process to save you time.
The kit consists of the following items:
  • Sampling probe to capillary sleeving (x2)
  • Nebulizer tubing with connector (x2)
  • Drain tubing with connector (x2)
  • 4mm connector for nebulizer gas
  • Sampling O-ring kit
  • Sampling probe (x2)
  • Capillary tubing (1.6mm OD x 0.8mm ID x 5000mm)
  • Nebulizer tubing with non-return valve
Specifications
For Use With (Equipment)iCAP 7000 Series ICP-OES Systems
TypeSample Introduction Spares Kit
Unit SizeEach