Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Thermo Scientific™
Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance plasma torch for iCAP™ Series ICP-OES systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
842312051841EMT Duo TorchiCAP 7000 Series
BRE0012328EMT Duo Plasma TorchiCAP PRO Series
842312051741EMT Radial TorchiCAP 7000/PRO Series
Catalog number 842312051841
Price (USD)
369.00
Each
Type:
EMT Duo Torch
For Use With (Equipment):
iCAP 7000 Series
Price (USD)
369.00
Each
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for Thermo Scientific™ iCAP™ Series ICP-OES systems. The torch clips in and can be removed in just seconds.
Enhanced performance through advanced accessories
We offer dedicated torch designs for radial and axial plasma views, including:
  • EMT Radial Torch for radial-only systems
  • EMT Duo Torch for radial and axial (duo) view systems
Specifications
For Use With (Equipment)iCAP 7000 Series
TypeEMT Duo Torch
Unit SizeEach