Pierce™ 1-Step Transfer Buffer

Catalog number: 84742

Thermo Scientific™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.

Features of 1-Step Transfer Buffer:

Fast—high ionic strength formulation provides 5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems
Ready to use—supplied as a 1X solution that is stable and ready to use at room temperature
Optimized—designed for seamless and reliable performance with the Pierce G2 Fast Blotter
Compatible—effective with other semi-dry transfer devices equipped with a suitable high-current power supply, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217)
Economical—excellent performance without the special or costly consumables that are required by other fast semi-dry transfer devices

When used with the Pierce G2 Fast Blotter, this buffer provides effective gel-to-membrane transfer of proteins in 5 to 10 minutes with efficiency that is equal to, or better than, conventional Western blotting techniques. Pierce 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217), when they are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.

Requires:
Pierce G2 Fast Blotter (Part No. 62288) or other semi-dry transfer device that is equipped or paired with a capable high-current power supply.

Fast blotting methods require a high-current power supply, such as the Pierce G2 Fast Blotter, and an optimized high ionic strength transfer buffer, such as Pierce 1-Step Transfer Buffer. By increasing the current, excellent transfer efficiency can be achieved in much shorter time compared to conventional methods. Amperage is held at a constant rate based on the surface area of the transfer stack(s) (~22-23mA/cm2) and voltage is limited to 25V.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: SDS-PAGE Gel (any)
Buffer Type: Transfer Buffers
Concentrated: 1 X
Gel Compatibility: SDS-PAGE Gels
Product Line: Pierce™
Product Size: 200 mL

Contents & storage

Upon receipt store at room temperature. Product is shipped at ambient temperature.

Documents

Manuals & protocols