How do I measure the digestion efficiency of my mass spec sample?
Digestion efficiency can be measured by assessing the number of missed cleaved peptides in the sample. The Pierce Digestion Indicator for Mass Spectrometry (Cat. No. 84841) can also be added to the sample to monitor digestion efficiency.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.