GeneArt™ Cloning into Customer Vector, 1 each - FAQs
GeneArt™ Cloning into Customer Vector, 1 each - FAQs
View additional product information for GeneArt Gene Synthesis design and ordering tool - FAQs (817005DE, 850310DE, 817000DE, 854005DE, 854004DE, 878016DE)
5 product FAQs found
What are the gene synthesis requirements for Express cloning?
Genes qualifying for Express cloning must be <4 kb and not complex (optimization of your sequence can reduce complexity).
What vectors are available for Express cloning?
The following vectors are currently available:
- pcDNA 3.1(+)vector
- pcDNA3.3-TOPO vector
- pcDNA3.4-TOPO vector
- pFastBac1 vector
- pET100/D-TOPO vector
- pET151/D-TOPO vector
- pRSET A vector
- pYes2.1V5-His TOPO vector
What will I receive if I use your GeneArt Express Cloning Service?
You should receive your synthesized gene in the selected expression vector. Please note, no additional copy in a pMx cloning vector will be provided as with traditional GeneArt Cloning Services.
What is the GeneArt Express Cloning Service?
Using an alternative production procedure with pre-prepared expression vectors, our GeneArt custom service team can clone your synthesized genes directly into selected Thermo Fisher Scientific expression plasmids, saving 4-5 business days compared to the standard workflow with cloning into a pMx series vector first. Please visit this page (http://www.thermofisher.com/us/en/home/life-science/cloning/gene-synthesis/geneart-plasmid-services/geneart-subcloning-service.html) for the differences between Express cloning and Classical cloning.
When should I use a GeneArt Gene Synthesis Kit, order GeneArt Strings DNA Fragments, or use GeneArt Gene Synthesis services?
The kit and custom services offer different advantages. Please click here (http://www.thermofisher.com/us/en/home/life-science/cloning/gene-synthesis/geneart-gene-synthesis-product-selection-guide.html) to view a selection guide comparing the technologies, including deliverables and production time, to determine what is best for you.