Histostain™-Plus 3rd Gen IHC Detection Kit - FAQs
Histostain™-Plus 3rd Gen IHC Detection Kit - FAQs
View additional product information for Histostain™-Plus 3rd Gen IHC Detection Kit - FAQs (859073)
1 product FAQs found
I am getting high DAB background staining. How can I decrease this background in the future?
High DAB background can be decreased by taking the following steps:
1) Make sure that the stringent wash is performed properly.
2) After hybridization, rinse the slides briefly in room temperature SSC buffer, then immerse slides for 5 minutes in SSC at 75°C.
3) Increase the temperature by 1°C per slide for more than 2 slides, but do not exceed 80°C.
4) For any other wash steps , 1X PBS with Tween 20 (0.025%) is recommended. Washing with any other reagents (1X PBS without Tween 20 or distilled water) can lead to unwanted background.