Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, Immulon, 4 HBX, High Flange, 120μL - FAQs
Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, Immulon, 4 HBX, High Flange, 120μL - FAQs
View additional product information for Clear Flat-Bottom Immuno Nonsterile 384-Well Plates - FAQs (8755)
1 product FAQs found
For Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, is the surface treatment solely on the bottom of the well, or is it also applied to the sides?
For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.