Subcellular Protein Fractionation Kit for Tissues - FAQs
Subcellular Protein Fractionation Kit for Tissues - FAQs
View additional product information for Subcellular Protein Fractionation Kit for Tissues - FAQs (87790)
3 product FAQs found
Can you provide the shelf-life for the Subcellular Protein Fractionation Kit for Tissues?
The Subcellular Protein Fractionation Kit for Tissues is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What kits do you offer for cell fractionation?
We offer the following kits for cell fractionation:
Mem-PER Plus Membrane Protein Extraction Kit, Cat. No. 89842
NE-PER Nuclear and Cytoplasmic Extraction Reagents, Cat. No. 78835
Subcellular Protein Fractionation Kit for Cultured Cells, Cat. No. 78840
Subcellular Protein Fractionation Kit for Tissues, Cat. No. 87790
Mitochondrial Isolation Kit for Cultured Cells, Cat. No. 89874
Mitochondrial Isolation kit for Tissue, Cat. No. 89801
Lysosome Enrichment Kit for Tissues and Cultured Cells, Cat. No. 89839
Find additional tips, troubleshooting help, and resources within our Cell Lysis and Fractionation Support Center.
Why is cell fractionation important?
Isolation of subcellular fractions and concentration of low-abundantce proteins allows for more efficient identification and study of the proteins of interest.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.