Pierce™ Tissue Strainers, 250 μm, 2.5 mL
Thermo Scientific™

Pierce™ Tissue Strainers, 250 μm, 2.5 mL

The Thermo Scientific Pierce Tissue Strainer fits standard 15 mL conical tubes for quick separation of cells or cell lysateRead more
Have Questions?
Catalog NumberQuantity
8779150 Units
Catalog number 87791
Price (USD)
152.65
Online exclusive
182.00
Save 29.35
Each
Quantity:
50 Units
Request bulk or custom format
Price (USD)
152.65
Online exclusive
182.00
Save 29.35
Each
The Thermo Scientific Pierce Tissue Strainer fits standard 15 mL conical tubes for quick separation of cells or cell lysate from tissue debris through a 250 μM mesh filter after mechanical disruption using gravity or gentle centrifugation.

Features of the Pierce Tissue Strainer:

• Convenient—remove tissue debris from tissue homogenate within minutes using gravity filtration or gentle centrifugation
• Compatible—unique design fits into standard 15 mL conical tubes
• Versatile—remove debris from a variety of tissues types, including liver, heart, brain, kidney, lung and spleen

The easy-to-use Pierce Tissue Strainers are made of strong nylon and enable rapid isolation cells or uniform protein extracts from up to 2.5 mL of tissue sample per load. The tissue strainers fit into standard 15 mL conical tubes, making the processing and recovery of samples simple and reliable.

Applications:
• Use with the Subcellular Protein Fractionation Kit for Tissues and other protein extraction reagent protocols that start with tissue samples.
• Improve uniformity of tissue homogenates to facilitate sample handling.
• Protect downstream instrumentation and devices by removing tissue clumps and debris that may foul injection ports and columns.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity50 Units
Product LinePierce
Product TypeTissue Strainer
Unit SizeEach
Contents & Storage
250μM Nylon Mesh on Inert Plastic Frame

Frequently asked questions (FAQs)

Can I autoclave Pierce Tissue Strainers, 250 µm, 2.5 mL (Cat. No. 87791)?

Pierce Tissue Strainers, 250 µm, 2.5 mL (Cat. No. 87791), can be autoclaved at 121 degrees C for 20-30 min without loss of function. Because they are made from nylon, they cannot withstand higher temperatures or longer durations.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.