Mouse IgG3 Uncoated ELISA Kit with Plates - FAQs
Mouse IgG3 Uncoated ELISA Kit with Plates - FAQs
View additional product information for Mouse IgG3 Uncoated ELISA Kit with Plates - FAQs (88-50440-86, 88-50440-76, 88-50440-22)
5 product FAQs found
What type of plates are included in the Ready-SET-Go! ELISA sets?
Corning Costar 96-well plates are included with Ready-SET-Go! ELISA catalog numbers ending in suffixes -22, -86 and -76. These solid 96-well plates are optional with the 10 and 20 plate kit sizes.
What is the sensitivity of your Ready-Set-Go! ELISA Sets?
The sensitivity ranges of our Ready-SET-Go! ELISA sets vary depending on the cytokine/protein being measured. Please refer to the product datasheet for set specific information.
What is the expiration date for your Ready-Set-Go! ELISA Sets?
The majority of our Ready-Set-Go! ELISA Sets are guaranteed for one year from the date of shipment. However, we do have several sets that are guaranteed for 6 months upon date of receipt. Please refer to the Certificate of Analysis for shelf-life information.
What are the storage conditions for your Ready-Set-Go! ELISA Sets?
All of the reagents are to be stored at 4 degrees C upon receipt, except for the recombinant standards. The recombinant standards MUST be stored at or below -80 degrees C to ensure stability and performance.
What do Ready-SET-Go! ELISA Sets contain?
Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant cytokine for generating standard curve and calibrating samples
10X Coating Buffer
5X ELISA/ELISPOT Diluent
Detection enzyme: pre-titrated Avidin-HRP
Substrate Solution: 1X TMB (Tetramethylbenzidine) Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018