81-Well Cryogenic Storage Racks with Lid
Thermo Scientific™
81-Well Cryogenic Storage Racks with Lid
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
8800Natural
8810Fluorescent Green
8820Fluorescent Orange
8830-11Fluorescent Pink
Catalog number 8800
Price (USD)
375.36
Online exclusive
408.00
Save 32.64
Case of 20
Color:
Natural
Price (USD)
375.36
Online exclusive
408.00
Save 32.64
Case of 20
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
Specifications
Holds81 (1.5 or 2.0 mL Tubes)
ColorNatural
MaterialPolypropylene
Product Type81-Well Cryogenic Storage Racks with Lid
Unit SizeCase of 20