Product Image
Invitrogen™
MagniSort™ Human CD56 Positive Selection Kit
The MagniSort™ Human CD56 Positive Selection Kit is designed for the magnetic separation of CD56+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6835-74200 tests
Catalog number 8802-6835-74
Price (USD)
745.65
Online Exclusive
994.00
Save 248.35 (25%)
Each
Quantity:
200 tests
Price (USD)
745.65
Online Exclusive
994.00
Save 248.35 (25%)
Each
The MagniSort™ Human CD56 Positive Selection Kit is designed for the magnetic separation of CD56+ cells by positive selection. It has been optimized for the isolation of CD56+ cells from human normal peripheral blood mononuclear cells utilizing a biotinylated Anti-human CD56 antibody and streptavidin-coated magnetic beads. CD56+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD56+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD56 (clone CMSSB).
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD56+
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypePositive Selection Kit
Unit SizeEach