Product Image
Invitrogen™
MagniSort™ Human CD4 Naïve T cell Enrichment Kit
The MagniSort™ Human CD4 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD4 naïve T cellsRead more
Have Questions?
Catalog NumberQuantity
8804-6814-74200 tests
Catalog number 8804-6814-74
Price (USD)
1,084.00
Each
Quantity:
200 tests
Price (USD)
1,084.00
Each
The MagniSort™ Human CD4 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD4 naïve T cells by negative selection. It has been optimized for the isolation of human CD4 naïve T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 naïve T cells untouched and free in solution.
The MagniSort™ Human CD4 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD8 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RO Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (Naïve, CD4)
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypeCell Enrichment Kit
Unit SizeEach
Frequently asked questions (FAQs)
I need to further purify my naive human T cells that have been isolated using the MagniSort Human CD4 Naive T cell Enrichment Kit (Cat. No. 8804-6814-74). Which purification kit can I use?
The CD4 naive T cells that are isolated using the MagniSort Human CD4 Naive T cell Enrichment Kit are untouched and free in solution. Hence, you can use any other purification kit afterwards.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.