Product Image
Invitrogen™

eBioscience™ BrdU Staining Kit for Flow Cytometry PE

The eBioscience BrdU Staining Kit for Flow Cytometry contains the necessary reagents and buffers for identifying and examining proliferating cellsRead more
Have Questions?
Catalog NumberQuantity
8812-6600-42100 tests
Catalog number 8812-6600-42
Price (USD)
718.65
Online exclusive
958.00
Save 239.35
Each
Quantity:
100 tests
Price (USD)
718.65
Online exclusive
958.00
Save 239.35
Each
The eBioscience BrdU Staining Kit for Flow Cytometry contains the necessary reagents and buffers for identifying and examining proliferating cells of mammalian species by flow cytometric analysis. Cycling cells are incubated with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis. Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest.

This kit is optimized to achieve brighter staining compared to traditional protocols.

Reactivity/Species
Bovine, Canine, Cow, Human, Monkey, Mouse, Non-human primate, Rat, Rhesus

Conjugate
PE

Laser
Blue Laser

Emit
575 nm

Excite
488 nm

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
FormatKit
Quantity100 tests
For Use With (Application)BrdU Staining
For Use With (Equipment)Flow Cytometer
Product LineeBioscience
Product TypeStaining Kit
Unit SizeEach

Frequently asked questions (FAQs)

Can I also stain for intracellular targets such as transcription factors and cytokines using the eBioscience BrdU staining kit?

Yes, primary antibodies against additional intracellular targets can be added alongside the Anti-BrdU antibody. However, it is critical that you empirically determine whether these additional antibodies are compatible with this staining protocol.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is the BrdU in the eBioscience BrdU staining kit supplied as a sterile formulation for in vivo use?

Yes, the BrdU is supplied as a sterile liquid. The vial should be opened under sterile conditions if it will be used in vivo.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I keep the diluted (1 mM) BrdU in the eBioscience BrdU staining kit at 4 degrees C? Can I refreeze it after use?

The stock (32.5 mM) BrdU can be frozen and thawed up to three times. Furthermore, the stock BrdU can be kept at 4 degrees C for up to one week. However, the diluted (1 mM) BrdU should be used immediately.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Have the BrdU Staining kits been tested on proliferating cells of different species?

We have used this kit to detect proliferating human and mouse cells. Other species have yet to be tested but feline, non-human primate and canine should also work.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the stopping points during flow cytometric analysis when using a BrdU staining kit?

Samples can be stored at 4 degrees C for up to 16 hours (i.e., overnight) after fixation with BrdU Staining Buffer (with or without washing in Flow Cytometry Staining Buffer) with no adverse effect on performance.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.