This product is only available through your local Thermo Scientific supplier. Learn where to buy >

pMCS Tluc16-DD Hygro Vector for Luciferase Assays

Catalog number: 88248

Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Thermo Scientific™ pMCS Tluc16-DD Hygro Vector is a multiple cloning site (MCS) vector designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc™16 (Tluc16) luciferase reporter.

• Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systems
• Multiple cloning sites provide versatility for transfer of regulatory elements from one vector to another
• Dual-destabilization (DD) technology reduces accumulation of Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assay
• Synthetic polyA terminator and Transcriptional Pause Site (TPS) included upstream of MCS to minimize non-specific transcriptional read-through
• Hygromycin resistance gene for use as selection marker to generate mammalian stable cell lines

Tluc16 luciferase is a 16 kDa, novel, intracellular luciferase derived from the marine copedod Metridia luciferase family. The wild-type luciferase has been modified to reduce its size, increase its brightness, and enable its intracellular expression.

The pMCS Tluc16-DD Hygro luciferase expression vector also contains dual-destabilization (DD) technology that reduces accumulation of the Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assay. A synthetic polyA terminator and a Transcriptional Pause Site (TPS) are included upstream of the MCS to minimize non-specific transcriptional read-through. This vector also contains the hygromycin resistance gene, which can be used as a mammalian selection marker to generate stable cell lines. The Tluc16 luciferase activity can be measured using the Thermo Scientific TurboLuc™ One-Step Glow Assay Kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Key Function: Reporter Assays
Reporter: TurboLuc™16 Luciferase
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Hygromycin
Product Line: Pierce™
Product Size: 10 µg

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols