Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL, 5 Columns - FAQs
Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL, 5 Columns - FAQs
View additional product information for Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL - FAQs (88277, 88276)
1 product FAQs found
Are Pierce High Capacity Endotoxin Removal Spin Columns, 1 mL (Cat. No. 88276) autoclavable?
We have not tested this, but the resin is autoclavable up to 121 degrees C at 1 bar for 20 min.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.