Pierce™ Primary Cardiomyocyte Isolation Kit - FAQs
Pierce™ Primary Cardiomyocyte Isolation Kit - FAQs
View additional product information for Pierce™ Primary Cardiomyocyte Isolation Kit - FAQs (88281)
1 product FAQs found
What kits are available for cell isolation?
We offer the following kits for cell isolation:
Primary Cardiomyocyte Isolation Kit, Cat. No. 88281
Mouse Embryonic Fibroblast Isolation Kit, Cat. No. 88279
Primary Neuron Isolation Kit, Cat. No. 88280
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.