Pierce™ Kinase Enrichment Kit with ATP Probe - FAQs
Pierce™ Kinase Enrichment Kit with ATP Probe - FAQs
View additional product information for Pierce™ Kinase Enrichment Kit with ATP Probe - FAQs (88310)
1 product FAQs found
Can I isolate my proteins based on their post-translational modification (PTM) or active binding site?
Enrichment of specific proteins or protein complexes can most easily be accomplished by using target-specific immobilized antibody or PTM specific affinity binding. We offer several kits targeting phosphorylated proteins, alpha- linked mannose and terminal glucose residues, N-acetyl glucosamine, polyubiquinated proteins, ATP binding sites, and active serine hydrolase enzymes. Choose the right protein enrichment kit for your application using the following link (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/protein-enrichment.html).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.