Pierce™ Triple Quadrupole Calibration Solution - FAQs
Pierce™ Triple Quadrupole Calibration Solution - FAQs
View additional product information for Pierce™ Triple Quadrupole Calibration Solution - FAQs (88325)
1 product FAQs found
Which instruments can be calibrated using the Pierce Triple Quadrupole Calibration Solution (Cat. No. 88325)?
The Pierce Triple Quadrupole Calibration Solution (Cat. No. 88325) is a mixture of high purity, ionizable components specifically designed for positive mode calibration of Thermo Scientific Triple Stage Quadrupole instruments such as TSQ Quantum, TSQ Discovery, TSQ Quantum Ultra, TSQ Quantum Access, TSQ Vantage, TSQ Endura, and TSQ Quantiva Series Mass Spectrometer instruments.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.