Pierce™ Reserpine Standard for LC-MS - FAQs
Pierce™ Reserpine Standard for LC-MS - FAQs
View additional product information for Pierce™ Reserpine Standard for LC-MS - FAQs (88326)
3 product FAQs found
What is the Pierce Reserpine Standard used for?
The Thermo Scientific Pierce Reserpine Standard for LCMS is a standard for determining the performance and sensitivity of all Thermo Scientific mass spectrometry instruments. This standard must be diluted before use according to the procedure specified in the corresponding instrument's Getting Started guide.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Is there a way to evaluate the sensitivity of my triple quad mass spectrometry instrument?
Yes, Pierce Reserpine Standard for LC-MS (Cat. No. 88326) can be used to assess instrument sensitivity.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
My spray stability is erratic in my mass spectrometer, can you help?
Yes, we offer the Pierce Reserpine Standard for LC-MS (Cat. No. 88326) to guide in troubleshooting spray stability.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.