4-Way Flipper™ Racks
Actual product may vary
Thermo Scientific™
4-Way Flipper™ Racks
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
8850Fluorescent Green
8860Fluorescent Orange
8870Fluorescent Pink
Catalog number 8850
Price (USD)
546.48
Online exclusive
594.00
Save 47.52
Case of 20
Color:
Fluorescent Green
Price (USD)
546.48
Online exclusive
594.00
Save 47.52
Case of 20
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.
Specifications
Description4-Way Flipper Rack
No. per Case20 Racks
ColorFluorescent Green
Holds0.5 mL, 15 mL, Tubes (50 mL)
Positions32 x 0.5 mL, 32 x 1.5 mL, 12 x 15 mL or 4 x 50 mL tubes, each on a different side
Product Type4-way Flipper Rack
ShapeRectangular
Unit SizeCase of 20