Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezer, 24.1 cu. ft. (681.5L) capacity, holds 500 boxes, 208-230V/60Hz - FAQs
Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezer, 24.1 cu. ft. (681.5L) capacity, holds 500 boxes, 208-230V/60Hz - FAQs
View additional product information for Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (88500D)