Pierce™ Protein Concentrators PES, 100K MWCO, 0.5–100 mL
Pierce™ Protein Concentrators PES, 100K MWCO, 0.5–100 mL
Pierce™ Protein Concentrators PES, 100K MWCO, 0.5–100 mL
Pierce™ Protein Concentrators PES, 100K MWCO, 0.5–100 mL
Thermo Scientific™

Pierce™ Protein Concentrators PES, 100K MWCO, 0.5–100 mL

Thermo Scientific Pierce Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of proteins over 100 KDa, in sample volumes from 0.5 to 100 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySample Volume
885374 Concentrators20 to 100 mL
8850325 Concentrators≤0.5 mL
8852310 Concentrators2 to 6 mL
8852424 Concentrators2 to 6 mL
8853210 Concentrators5 to 20 mL
8853324 Concentrators5 to 20 mL
Catalog number 88537
Price (USD)
194.65
Online exclusive
216.00
Save 21.35
Each
Quantity:
4 Concentrators
Sample Volume:
20 to 100 mL
Price (USD)
194.65
Online exclusive
216.00
Save 21.35
Each
Easily and quickly concentrate 100K molecular weight cutoff (MWCO) protein and biological samples with Thermo Scientific Pierce Protein Concentrators PES (polyethersulfone), which are disposable centrifugation devices that enable fast sample recovery and a short processing time. These concentrator columns can be used with standard microcentrifuge equipment, and samples can be collected without the need for invert spinning. Samples can be concentrated with or without desalting.

For protein and other biological samples, such as nucleic acids, with a 100K MWCO, Pierce Protein Concentrators PES offer rapid protein concentration, excellent sample recovery, and instrument compatibility. These centrifugal devices feature a high-flux polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples. This product has a MWCO of 100K and can be used for processing sample volumes of 0.5–100 mL.

Features of Protein Concentrators PES include:
• Rapid protein concentration—sample concentration of 10- to 30-fold can be achieved in 15 or fewer minutes
• Excellent protein recovery—typical protein recovery is >90% (when the sample mass is at least two-fold greater than the posted MWCO)
• Instrument-compatibility—a standard microcentrifuge with swinging or fixed-angle (except 88537) rotors can be used; sample can be collected and recovered without invert spinning
• Versatile—easily perform sample concentration, diafiltration, or buffer-exchange of biological samples

For easy identification, the MWCO value is etched on the side of each device. To assist with sample addition and recovery, a clear window with graduations is located on the side of each device enabling visual determination of the retentate volume. The vertical design of the membrane allows high-flux rates, low non-specific binding, and negligible protein aggregation on the membrane and provides reliable and consistent results for processing sample volumes between 0.5 and 100 mL. Typical protein recovery is 90% when the mass of the proteins in the sample is at least two-fold greater than the MWCO of the device used. For example, when using IgG (MW ∼150K) samples, use the 5K, 10K, 30K, or 50K MWCO concentrators. Sample concentration of 10- to 30-fold can be achieved within 15 minutes.

Protein Concentrators PES applications
• Protein concentration of tissue culture media, antiserum, or monoclonal antibody preparations
• Concentration of protein peaks following gel-permeation chromatography
• Removal of unincorporated label following protein modification reactions
• Concentration and desalt/buffer-exchange after elution of protein from ion-exchange, hydrophobic interaction (HIC), metal chelate, or affinity chromatography columns
• Separation of different sized molecules

Pierce Protein Concentrators PES contain a membrane rated for retaining molecules with a molecular weight at least two-fold greater than the MWCO rating of the PES membrane within the device. Reduced recovery may occur when using a concentrator with molecules smaller than the recommended mass. Protein recovery will vary depending on the specific protein in the sample and its starting concentration.

The dead-stop volume for the 0.5 mL, 2–6 mL, 5–20 mL, and 20–100 mL concentrators is approximately 15 μL, 30 μL, 50 μL, and 350 μL, respectively.

To achieve >90% recovery of protein with the 0.5 mL concentrators, the minimum protein sample concentration should be 0.05 mg/mL. For all other concentrators, the minimum protein sample concentration should be 0.1 mg/mL.

Optimal centrifuges and speeds
The 0.5 mL concentrators can be used effectively at a maximum centrifugal force of 15,000 × g using a fixed angle centrifuge only.

For the 2-6 mL concentrators, swinging-bucket or fixed-angle centrifuge models can be used. Swinging bucket centrifuges capable of generating 2,000–4,000 × g and a maximum centrifugal force of 4,000 × g are recommended. With fixed-angle centrifuges, models capable of generating 5,000–10,000 × g, and a maximum centrifugal force of 8,000 × g, are recommended.

For the 5-20 mL volume concentrators, swinging-bucket or fixed-angle centrifuge models can be used. Swinging-bucket centrifuges capable of generating 2,000–5,000 × g and a maximum centrifugal force of 4,000 × g are recommended. With fixed-angle centrifuges, models capable of generating 5,000–8,000 × g and a maximum centrifugal force of 6,000 × g are recommended.

For the 20-100 mL concentrators, only swinging-bucket centrifuges capable of generating 1,000–2,000 × g are recommended. Use a maximum centrifugal force of 2,000 × g. These concentrators are not recommended for use in fixed-angle rotors.

Centrifugal force, temperature, sample volume, concentration, and viscosity will all affect filtration rate. Centrifugation time should therefore be optimized for each application.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LinePierce
Purification TargetProtein
Quantity4 Concentrators
Sample Volume20 to 100 mL
Shipping ConditionRoom Temperature
TypeFilter
DisposableSingle-use
FormatBottle, Spin Column
MWCO100 kDa
Material (Membrane)PES
Unit SizeEach
Contents & Storage
Store at Room Temperature.

Frequently asked questions (FAQs)

Besides precipitation with ammonium sulfate, what are some ways I can concentrate my protein sample?

You may remove excess solvent and smaller moieties by centrifugation through a microconcentrator. This will concentrate your protein sample.

(1) Choose microconcentrator tube (available from a variety of commercial suppliers) with a protein cutoff smaller than the molecular weight of the protein in the sample. Check our Pierce Protein Concentrators PES.

(2) Add 1 µL of 20% w/v SDS to a dry microconcentrator tube (if sample does not already contain SDS).

(3) Slowly add sample (a few microliters at a time) to membrane until membrane is completely wet. Centrifuge to near (but not complete) dryness.

(4) If intention is to desalt sample or buffer exchange: Add ~50 µL water to microconcentrator and spin until nearly dry. Repeat buffer exchange. Sample will remain on membrane. Check our Zeba desalting proteins.

(5) Using a new collection tube, invert membrane and spin at low speed (1000 x g) to elute protein from membrane. Add 2X SDS-Sample Buffer containing 10 mM DTT to membrane: vortex, invert and spin. Final volume should be ~20 µL.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the dimensions of Pierce Protein Concentrators PES, 100K MWCO, 0.5–100 mL (Cat. No. 88537)?

The dimensions are:
61 mm in outer diameter
124 mm in length

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What bucket adaptors are recommended for 20-100 mL capacity Pierce Protein Concentrators PES?

The dimensions of the 20-100 mL capacity concentrators (Pierce Protein Concentrators PES, 3K or 5K MWCO, 0.5–100 mL (Cat. No. 88534), Pierce Protein Concentrators PES, 10K MWCO, 0.5–100 mL (Cat. No. 88535), Pierce Protein Concentrators PES, 30K MWCO (Cat No. 88536), Pierce Protein Concentrators PES, 50K MWCO, 0.5–100 mL (Cat. No. 88542), and Pierce Protein Concentrators PES, 100K MWCO, 0.5–100 mL (Cat No. 88537)) are 61 mm outer diameter and 124 mm height. These are similar to Nalgene and Nunc 250 mL bottles. Adapters to the Nunc 250 mL bottles will fit to the 20-100 mL capacity concentrators.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What kind of rotor do you recommend using with a protein concentrator that can handle 20-100 mL sample volume?

We recommend using a rotor that can accommodate a 250 mL Nalgene bottle.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can I autoclave PES protein concentrators?

We do not recommend autoclaving the PES protein concentrators. High temperatures will significantly increase the membrane molecular weight cutoff (MWCO). To sterilize, we recommend using a 70% ethanol solution.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.