Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1L) capacity, holds 600 boxes, 208-230V/60Hz - FAQs
Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1L) capacity, holds 600 boxes, 208-230V/60Hz - FAQs
View additional product information for Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (88600D)