Pierce™ Direct Magnetic IP/Co-IP Kit - FAQs
Pierce™ Direct Magnetic IP/Co-IP Kit - FAQs
View additional product information for Pierce™ Direct Magnetic IP/Co-IP Kit - FAQs (88828)
2 product FAQs found
Can you provide the shelf-life for Direct Magnetic IP/Co-IP Kit?
Direct Magnetic IP/Co-IP Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
I am using the Pierce Direct Magnetic IP/Co-IP Kit. The magnetic beads formed an aggregate during the coupling process; how do I avoid this?
The proteins on the bead surface may adhere to the plastic tube and to each other. To avoid this, after the quenching step, add a mild surfactant at a final concentration of 0.05% (v/v) ( e.g., Tween-20 Detergent, Cat. No 85113) to the washes
Note: Do not use detergent in the coupling step.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.