HisPur™ Ni-NTA Magnetic Beads
Thermo Scientific™

HisPur™ Ni-NTA Magnetic Beads

Perform affinity purification of His-tagged fusion proteins in manual or automated formats with the high capacity nickel-IMAC Thermo Scientific™ HisPur Ni-NTA Magnetic Beads.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
888312 mL
8883210 mL
Número do catálogo 88831
Preço (BRL)
2.485,34
Each
Quantity:
2 mL
Request bulk or custom format
Preço (BRL)
2.485,34
Each

Thermo Scientific HisPur Ni-NTA Magnetic Beads are high-capacity nickel-IMAC beads for affinity purification of His-tagged fusion proteins in manual or automated formats.

Features of HisPur Ni-NTA Magnetic Beads:

  • High capacity—equivalent or higher binding capacity than Ni-NTA magnetic beads from other suppliers
  • Low nonspecific binding—the bead surface is pre-blocked and the protocol provides optimized buffers for purification
  • Fast—protocol is completed in 1 hour
  • Scalable—process microliter to milliliter sample volumes
  • Versatile—purify proteins using native or denaturing conditions
  • Reagent compatible—can be used with common cell lysis reagents and a variety of buffer additives
  • Multiple formats—protein coupling to the beads and downstream applications can be performed both manually and on an automated platform (e.g., Thermo Scientific KingFisher Instruments)

The blocked magnetic bead surface is derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) beads provide high binding capacity with very low background. The HisPur Ni-NTA Magnetic Beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments for high-throughput needs.

HisPur Ni-NTA Magnetic Beads are used for small scale affinity purification as well as high-throughput screening of recombinant His-tagged proteins. The polyhistidine tag is the most popular affinity tag and typically consists of six consecutive histidine residues (6xHis). These tagged proteins are overexpressed in a number of different systems, most commonly in bacteria, and purified from cell lysates such as those prepared using B-PER Bacterial Protein Extraction Reagents. Purification of His-tagged proteins is achieved using a NTA chelate charged with nickel that coordinates with the histidine side chains. The NTA chelate contains four metal-binding sites which allow for low metal ion leaching and high binding capacity. The protocol for the HisPur Ni-NTA Magnetic Beads has been optimized to allow for high purity of the isolated His-tagged protein. Performance is equivalent to or better than Ni-NTA magnetic beads from other suppliers.

For Research Use Only. Not for use in diagnostic procedures.
Especificações
Capacity (Metric)2 mL
ConcentrationSlurry: 12.5mg/mL, 1.25% solids
Ligand TypeNickel-NTA
Protein FormRecombinant
Quantity2 mL
TargetHis
FormLiquid
Particle Size1 μm
Product LineHisPur
TypeMagnetic Affinity Bead
Unit SizeEach
Conteúdo e armazenamento
Store at 4°C

Frequently asked questions (FAQs)

Are there other sequences that can bind to nickel more tightly than 6xHis-tagged proteins and how can they be eluted.

Yes, 7xHis-tagged proteins, proteins naturally high in histidine, and other combinations of His and other amino acids will bind. To elute them, you have to increase the concentration of imidazole. Generally these peptides will not contaminate your fraction since they remain on the column. However after multiple uses of the same column, these peptides may reduce the binding capacity of the column.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What should the typical protein recovery be when using the Probond Purification System or Ni-NTA Purification system?

Both systems are qualified by purifying 2 mg of myoglobin protein on a column and performing a Bradford assay. Protein recovery must be 75% or higher.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the difference between HisPur Nickel Resin and HisPur Cobalt Resin?

Nickel has a higher affinity for histidines than does cobalt. It binds multiple histidines more tightly than cobalt, and requires more stringent conditions than is necessary for cobalt. The lower affinity of cobalt for multiple histidines typically results in less nonspecific binding of histidine-rich proteins that lack a his-tag compared to nickel resins.

HisPur Ni-NTA resin has a high capacity of up to 60 mg of 6xHis-tagged protein per milliliter. It is a versatile resin that can work under both native and denaturing conditions, and can also be used with a variety of lysis reagents and buffer additives.

HisPur Cobalt resin utilizes proprietary tetradentate chelating resin charged with cobalt. This system recovers highly purified protein with lower imidazole concentrations and has low metal leeching properties.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citações e referências (6)

Citações e referências
Abstract
Chapter 1 - Isolation of mitochondria from cells and tissues
Authors:Pin-Chao Liao, Christian Bergamini, Romana Fato Liza A.Pon Francesco Pallotti
Journal:Methods in Cell Biology
PubMed ID:
Neuronal death in pneumococcal meningitis is triggered by pneumolysin and RrgA interactions with b-actin
Authors:Tabusi M, Thorsdottir S, Lysandrou M, Narciso AR, Minoia M, Srambickal CV, Widengren J, Henriques-Normark B, Lovino F
Journal:PLOS Pathogens
PubMed ID:
Diverse high-affinity DNA aptamers for wild-type and B.11.7 SARS-CoV-2 spike proteins from a pre-structured DNA library
Authors:Li J, Zhang Z, Gu J, Stacey HD, Ang JC, Capretta A, Filipe CDM, Mossman KL, Balion C, Salena BJ, Yamamura D, Soleymani L, Miller MS, Brennan JD, Li Y
Journal:Nucleic Acids Research Oxford Academic (oup.com)
PubMed ID:
Neuronal death in pneumococcal meningitis is triggered by pneumolysin and RrgA interactions with b-actin
Authors:Tabusi M, Thorsdottir S, Lysandrou M, Narciso AR, Minoia M, Srambickal CV, Widengren J, Henriques-Normark B, Lovino F
Journal:PLOS Pathogens
PubMed ID:
Chapter 1 - Isolation of mitochondria from cells and tissues
Authors:Pin-Chao Liao, Christian Bergamini, Romana Fato Liza A.Pon Francesco Pallotti
Journal:Methods in Cell Biology
PubMed ID: