pT7CFE1-CHis Vector for Mammalian Cell-Free Protein Expression - FAQs
pT7CFE1-CHis Vector for Mammalian Cell-Free Protein Expression - FAQs
View additional product information for pT7CFE1-CHis Vector for Mammalian Cell-Free Protein Expression - FAQs (88860)
1 product FAQs found
How is the stock vector supplied in pT7CFE1-CHis Vector for Mammalian Cell-Free Protein Expression (Cat. No. 88860)?
10 µg of the pT7CFE1-CHis vector is supplied at a concentration of 500 ng/µL.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.