pT7CFE1-CGFP-HA-His Vector for Mammalian Cell-Free Protein Expression
Thermo Scientific™
pT7CFE1-CGFP-HA-His Vector for Mammalian Cell-Free Protein Expression
Thermo Scientific pT7CFE1-CGFP-HA-His is a cloning plasmid optimized to use with the Thermo Scientific 1-Step Human In Vitro Protein ExpressionRead more
Have Questions?
Catalog NumberQuantity
8886910 μg
Catalog number 88869
Price (USD)
137.00
Each
Quantity:
10 μg
Request bulk or custom format
Price (USD)
137.00
Each
Thermo Scientific pT7CFE1-CGFP-HA-His is a cloning plasmid optimized to use with the Thermo Scientific 1-Step Human In Vitro Protein Expression System for in vitro translation (IVT) of tagged fusion proteins. pT7CFE1-CGST-HA-His Vector is available with tandem affinity tags, GFP,HA and 6xHis at the C-terminus, to facilitate protein purification and detection. pT7CFE1-CGST-HA-His also has a cleavable tag, HRV 3C, available on the C-terminus.
Features of pT7CFE1:
• EMCV IRES at the 5' UTR promotes high-level translation of mRNAs
• MCS accommodates gene insertion via ten different restriction sites: Msc1, Nde1, BamH1, EcoR1, EcoRV, Pac1, Pst1, Sac1, Sal1, Not1 and Xho1
• Poly A sequence in the 3' region promotes mRNA stabilization and protection from nucleases
• T7 terminator ensures synthesis of accurate size mRNA transcripts
• Plasmid linearization may be accomplished with restriction sites between Poly A sequence and the T7 terminator region
pT7CFE1 Expression Vectors contain the Encephalomyocarditis virus (EMCV) internal ribosome entry site (IRES) element that is critical for high levels of cap-independent protein expression in the Human In Vitro Translation System. Each vector features a highly-compatible multiple cloning site (MCS) to facilitate easy insertion of protein coding sequences into and between vectors. The pT7CFE1 Vector is available with single or tandem affinity tags at the N- or C- terminus to facilitate protein purification and detection. The pT7CFE Vectors are suitable for insertion of cloned genes, cDNAs, ORFs or PCR products for in vitro transcription and translation. Custom cloning services are also available.
More Product Data
• Choosing a vector and purification method for in vitro protein expression
Related Products
pT7CFE1-NHA-CHis Vector for Mammalian Cell-Free Protein Expression
pT7CFE1-CGST-HA-His Vector for Mammalian Cell-Free Protein Expression
pT7CFE1-NHis-GST Vector for Mammalian Cell-Free Protein Expression
pT7CFE1-NHis-GST-CHA Vector for Mammalian Cell-Free Protein Expression
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Cloning MethodRestriction Enzyme/MCS
DescriptionpT7CFF1-CGFP-HA-His Vector
Protein TagHis Tag, turboGFP, HA Tag
Quantity10 μg
PromoterT7
TypepT7CFE1-CGFP-HA-His Vector
Unit SizeEach
Contents & Storage
Store between -70°C and -20°C.