Touch Screen Dry Bath/Block Heater
Thermo Scientific™

Touch Screen Dry Bath/Block Heater

Achieve precise temperature stability and uniformity with our easy-to-use touch screen dry bath.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsPlug Type
888700101 BlockCN, EU, UK
888700071 BlockUS
888700082 BlocksUS
888700112 BlocksCN, EU, UK
888700094 BlocksUS
888700124 BlocksCN, EU, UK
Catalog number 88870010
Price (EUR)
621,54
Online exclusive
674,00
Save 52,46 (8%)
Each
Holds:
1 Block
Plug Type:
CN, EU, UK
Price (EUR)
621,54
Online exclusive
674,00
Save 52,46 (8%)
Each

Designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen controller. With 1, 2 and 4 block models, a broad range of heat blocks and a wide temperature range of ambient +5°C to 130°C, you're sure to find the dry bath to fit your application demands. The Thermo Scientific™ Touch Screen Dry Bath/Block Heater includes advanced features such as a timer, programmability with the ability to save up to 10 programs with 5 steps per program, and an optional external temperature sensor to control the heat setting by a real time sample read out.

  • Advanced internal temperature sensing probe with outstanding temperature accuracy and control
  • Temperature deviation adjustment
  • Precise temperature control with PID circuit
  • Temperature self-calibration enables user to offset the temperature to desired value
  • Timer enables the user to accurately monitor the heating time
  • Password protection feature
  • Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting
  • Range of configuration to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications
  • Powder-coated steel body construction ensures durability
  • When the external temperature sensor is connected, the internal temperature sensor will disconnect automatically, allowing the temperature control and display to rely on the external temperature sensor
  • Easy-to-use touch screen control displays time and temperature
  • 10 user setting programs, each supports up to 5 sequential steps of different temperatures and duration times
  • Built-in over temperature protection device for added safety

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation

Required accessory:

  • Unit must be ordered with block(s)
Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, cULus, RoHS
Controller TypePID digital
Dimensions (L x W x H)373 x 200 x 100 mm
Frequency50/60 Hz
Fuse250V 2.5A
Heating Rate≤ 20 min. 30° to 130°C
Height (English)3.9 in.
Height (Metric)100 mm
Holds1 Block
IncludesDry Bath, Block Lifter
Length (English)14.7 in.
Length (Metric)373 mm
LightingLED
No. of Blocks1 (sold separately)
Plug TypeCN, EU, UK
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 1°C
Timer0 to 99:59 min or continuous
TypeDry Block Heater
Warranty2 Year, Replacement
Weight (English)7.5 lb.
Weight (Metric)3.4 kg
Width (English)7.9 in.
Width (Metric)200 mm
MaterialPowder-coated Steel
Relative Humidity<80%
Temperature Range (Metric)Ambient +5° to 130°C (Ambient at 25°C)
Voltage200/240 V
Unit SizeEach