Touch Screen Dry Bath/Block Heater
Touch Screen Dry Bath/Block Heater
Touch Screen Dry Bath/Block Heater
Thermo Scientific™

Touch Screen Dry Bath/Block Heater

Achieve precise temperature stability and uniformity with our easy-to-use touch screen dry bath.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호보유플러그 유형
888700112 BlocksCN, EU, UK
888700071 BlockUS
888700101 BlockCN, EU, UK
888700082 BlocksUS
888700094 BlocksUS
888700124 BlocksCN, EU, UK
카탈로그 번호 88870011
제품 가격(KRW)
1,116,000
온라인 행사
Ends: 30-Sep-2026
1,312,000
할인액 196,000 (15%)
Each
보유:
2 Blocks
플러그 유형:
CN, EU, UK
제품 가격(KRW)
1,116,000
온라인 행사
Ends: 30-Sep-2026
1,312,000
할인액 196,000 (15%)
Each

Designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen controller. With 1, 2 and 4 block models, a broad range of heat blocks and a wide temperature range of ambient +5°C to 130°C, you're sure to find the dry bath to fit your application demands. The Thermo Scientific™ Touch Screen Dry Bath/Block Heater includes advanced features such as a timer, programmability with the ability to save up to 10 programs with 5 steps per program, and an optional external temperature sensor to control the heat setting by a real time sample read out.

  • Advanced internal temperature sensing probe with outstanding temperature accuracy and control
  • Temperature deviation adjustment
  • Precise temperature control with PID circuit
  • Temperature self-calibration enables user to offset the temperature to desired value
  • Timer enables the user to accurately monitor the heating time
  • Password protection feature
  • Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting
  • Range of configuration to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications
  • Powder-coated steel body construction ensures durability
  • When the external temperature sensor is connected, the internal temperature sensor will disconnect automatically, allowing the temperature control and display to rely on the external temperature sensor
  • Easy-to-use touch screen control displays time and temperature
  • 10 user setting programs, each supports up to 5 sequential steps of different temperatures and duration times
  • Built-in over temperature protection device for added safety

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation

Required accessory:

  • Unit must be ordered with block(s)
Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
사양
인증/적합성CE, cULus, RoHS
제어기 유형PID digital
치수(길이 x 폭 x 높이)403 x 200 x 100 mm
주파수50/60 Hz
퓨즈250V 2.5A
가열 속도≤ 20 min. 30° to 130°C
높이(영국식 단위)3.9 in.
높이(미터법)100 mm
보유2 Blocks
포함Dry Bath, Block Lifter
길이(영국식 단위)15.9 in.
길이(미터법)403 mm
조명LED
블럭 수2 (sold separately)
플러그 유형CN, EU, UK
온도 정확도≤ ± 0.5°C
온도 균일성≤ ± 1°C
타이머0 to 99:59 min or continuous
유형Dry Block Heater
워런티2 Year, Replacement
중량(영국식 단위)8.16 lb.
중량(미터법)3.7 kg
폭(영국식 단위)7.9 in.
폭(미터법)200 mm
물질Powder-coated Steel
상대 습도<80%
온도 범위(미터법)Ambient +5° to 130°C (Ambient at 25°C)
전압200/240 V
Unit SizeEach