Block, 1 x 96-well Elisa
Thermo Scientific™

Block, 1 x 96-well Elisa

For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog NumberNo. of Places
8887011196
Catalog number 88870111
Price (USD)
112.20
Online exclusive
132.00
Save 19.80
Each
No. of Places:
96
Price (USD)
112.20
Online exclusive
132.00
Save 19.80
Each
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
Description1 x 96-well Elisa block
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds1 x 96-well Elisa
LightingLED
TypeBlock
No. of Places96
Unit SizeEach