96-well Non-Skirted PCR Plate Block
Thermo Scientific™

96-well Non-Skirted PCR Plate Block

For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog NumberDimensions (L x W x H)
8887012076 x 124 x 39 mm
Catalog number 88870120
Price (USD)
289.00
Online exclusive
340.00
Save 51.00
Each
Dimensions (L x W x H):
76 x 124 x 39 mm
Price (USD)
289.00
Online exclusive
340.00
Save 51.00
Each
Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Required Accessory:
  • Unit must be ordered with block(s)
Specifications
DescriptionNon-skirted PCR Plate Block, 96 well
Dimensions (L x W x H)76 x 124 x 39 mm
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds1 x 96 well non-skirted PCR plate
LightingLED
TypeBlock
Capacity (Metric)0.2 mL
MaterialSteel
No. of Places96
Temperature Range (Metric)130°C
Unit SizeEach