Compact Digital Dry Bath/Block Heater
Thermo Scientific™

Compact Digital Dry Bath/Block Heater

Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelPlug Type
88871002Compact Dry Bath DUS
88871001Compact Dry Bath SUS
88871003Compact Dry Bath SCN, EU, UK
88871004Compact Dry Bath DCN, EU, UK
Catalog number 88871002
Price (USD)
645.15
Online exclusive
759.00
Save 113.85
Each
Model:
Compact Dry Bath D
Plug Type:
US
Price (USD)
645.15
Online exclusive
759.00
Save 113.85
Each
Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.
  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification

Required Accessory:

  • Unit must be ordered with block(s)
Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, cTUVus, RoHS
Controller TypeDigital
Dimensions (L x W x H)200 x 200 x 25 mm (7.9 x 7.9 x 1 in.)
Electrical Requirements12 V, DC, 7 A
Heating Rate20 min. average heating rate (30° to 100°C, ambient at 25°C)
Height (English)1 in.
Height (Metric)25 mm
IncludesDry Bath, US style plug, block lifter, plastic lid
Length (English)7.9 in.
Length (Metric)200 mm
LightingLED
ModelCompact Dry Bath D
No. of Blocks2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks
Plug TypeUS
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 0.5°C
TypeDry Block Heater
Warranty1 Year, Replacement
Weight (English)3.3 lb.
Weight (Metric)<1.5 kg
Width (English)7.9 in.
Width (Metric)200 mm
Capacity (Metric)0.2 mL
Temperature Range (Metric)Ambient +2° to 100°C
Voltage100/240 V
Unit SizeEach