tGFP mRNA for 1-Step Human IVT Kits
Thermo Scientific™
tGFP mRNA for 1-Step Human IVT Kits
tGFP mRNA (0.75 μg/μL in 0.1 mM EDTA) is intended as a control mRNA for use in in vitro proteinRead more
Have Questions?
Catalog NumberQuantity
8888010 μg
Catalog number 88880
Price (USD)
178.00
Each
Quantity:
10 μg
Request bulk or custom format
Price (USD)
178.00
Each
tGFP mRNA (0.75 μg/μL in 0.1 mM EDTA) is intended as a control mRNA for use in in vitro protein expression kits, such as the Thermo Scientific Human In Vitro Protein Expression Kit.
More Product Data
• Protocol for in vitro protein expression using mRNA templates
• Glycoprotein expression in a human IVT system
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce tGFP mRNA
Expressed Protein TypeMembrane Protein
FormatTube
Kit ContentsControl Template(s)
Label TypeUnconjugated
Product Line1-Step
Quantity10 μg
TargetGFP (Green fluorescent protein)
TypetGFP mRNA
Unit SizeEach
Contents & Storage
Store below -70°C.