You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Hotplates and Stirrers
▸
Stirrer Accessories
▸
Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers
Actual product may vary
Thermo Scientific™
Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers
Use these accessories to optimize performance of your Thermo Scientific™ RT Basic Series Magnetic Stirrers.
Have Questions?
Change view
Catalog Number
Diameter (Metric)
Type
88880144
120 mm
Plate Cover
88880145
170 mm
Plate Cover
88880146
220 mm
Plate Cover
3 Options
Catalog number
88880144
Price (USD)
-
Request A Quote
Diameter (Metric):
120 mm
Type:
Plate Cover
Use these Thermo Scientific™ Accessories for RT Basic Series Magnetic Stirrers to optimize performance of your stirrer.
Specifications
Color
White
Diameter (Metric)
120 mm
For Use With (Equipment)
RT Basic Series and RT Touch Series Magnetic Stirrers
Holds
96-Well Plates, 384-Well Plates
Material
Silicone
Type
Plate Cover
Unit Size
Each