Clamps for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Thermo Scientific™
Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeClamps
88881111Flask Clamp1000 mL
88881105Flask Clamp50 mL
88881135CFlask Clamp25 mL
88881106Flask Clamp100 mL
88881107Flask Clamp150 mL
88881108Flask Clamp200 mL
88881109Flask Clamp250 mL
88881110Flask Clamp500 mL
88881112Flask Clamp2000 mL
88881113Flask Clamp3000 mL
88881114Flask Clamp5000 mL
88881115Infusion bottle clamp500 mL
88881116Infusion bottle clamp1000 mL
88881117Funnel ClampSeparatory
88881133Starter KitFlask
Catalog number 88881111
Price (USD)
63.54
Online exclusive
70.60
Save 7.06
Each
Product Type:
Flask Clamp
Clamps:
1000 mL
Price (USD)
63.54
Online exclusive
70.60
Save 7.06
Each
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Clamps1000 mL
For Use With (Equipment)CO2 Resistant Shaker
No. of Positions6
Product TypeFlask Clamp
Unit SizeEach