Digital Cel-Gro Tissue Culture Rotators
Digital Cel-Gro Tissue Culture Rotators
Digital Cel-Gro Tissue Culture Rotators
Thermo Scientific™

Digital Cel-Gro Tissue Culture Rotators

The Digital Cel-Gro Tissue Culture Rotator has a brushless no-maintenance motor that provides a gentle rotating motion for optimum results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug TypeCertifications/Compliance
88882015
also known as 88882015-AVL
US PlugcCSAus
88882016
also known as 88882016-AVL
EU, UK, CN PlugCE
Catalog number 88882015
also known as 88882015-AVL
Price (USD)
2,074.00
Online exclusive
2,440.00
Save 366.00
Each
Plug Type:
US Plug
Certifications/Compliance:
cCSAus
Price (USD)
2,074.00
Online exclusive
2,440.00
Save 366.00
Each

The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.

  • Digital display of Speed and Time
  • Speed range: 5 – 70 rpm
  • Speed Accuracy: ±1 rpm
  • Adjustable rocking Angle: 95°- 180°
  • Loading Capacity: 5 kg
  • Timer Range: 0 - 9999mins
  • Optional multiple size drums offering different capacity of tube rotation
Specifications
AlarmsAudible alarm
Amperage0.2 A
Controller TypeDigital control
Dimensions (L x W x H)383 x 260 x 522 mm (15.1 x 10.2 x 20.6 in.)
DisplayDigital
Electrical Requirements100/240 V
For Use With (Application)Cell Culture
For Use With (Equipment)Accessories: 88882149, 88882150,88882151, 88882152, 88882153, 88882154, 88882155, 88882156, 88882157
Frequency50/60 Hz
Humidity Range20 to 85%
IncludesEquipment only
MaterialStainless Steel Platform
Plug TypeUS Plug
Shipping Weight (English)26 lb.
Shipping Weight (Metric)11.8 kg
Speed Control Accuracy±1 rpm
Tilt Angle95 to 180°
Timer Range0 to 9999 min.
Warranty and Service Offering2 Years
Certifications/CompliancecCSAus
Load Bearing Capacity (Metric)5 kg
ModelDigital Cel-Gro Tissue Culture Rotator
Speed Range5 to 70 rpm
Temperature Range (Metric)5°C to 40°C
Unit SizeEach